- Description:Urea Nitrogen (BUN) Colorimetric Detection Kit
- Size:2 Plate
- Cat. No.:MSPP-K024H1
The DetectX® urea nitrogen (BUN) detection kit is designed to quantitatively measure urea nitrogen in a variety of samples.
- Use - measure urea nitrogen in 30 minutes
- Sample - serum, plasma, urine, saliva and TCM
- Samples/kit - 88 or 472 in duplicate
- Calibrated - N-Cal kit, NIST-calibrated
- Stability - stable room temperature liquid reagents
A urea nitrogen standard calibrated to NIST reference materials is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Samples are mixed with Color reagents A and B and incubated at room temperature for 30 minutes. The colored product is read at 450 nm. The concentration of urea nitrogen in the sample is calculated, after making a suitable correction for any dilution, using software available with most plate readers. The results are expressed in terms of mg/dl urea nitrogen. If samples are to be expressed in terms of mg/dl urea, the data can be converted using the multiplier 2.14.