Order Entry
United States
ContactUsLinkComponent
Octet® Protein A (ProA) Biosensor, Sartorius
Octet® Protein A (ProA) Biosensor, Sartorius
Catalog # MSPP-185012
CAS Number:  
Octet® Protein A (ProA) Biosensor, Sartorius
Catalog # MSPP-185012
Supplier Number:  18-5012
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Protein A (ProA) biosensor, Five trays of 96 biosensors coated with Protein A (recommended Sample Diluent sold separately)
  • Cat. No.:
    MSPP-185012
  • Supplier no.:
    18-5012

Specifications

About this item

For determination of antibody concentration.

  • Direct measurement of immunoglobulins (IgG)
  • Assay samples without centrifugation
  • Fast turnaround of results
  • Broad dynamic range
  • Correlates to HPLC
  • Dynamic range 1–500 μg/ml for most proteins

Throughput: 8 samples in about 2 minutes, 96 samples in about 32 minutes.

Ordering information: Additional materials are required to run these assays. Recommended sample diluent sold separately. Standards: A purified standard that is identical to the experimental samples is required. Media for sensor hydration: the biosensor hydration solution must match the standard and experimental sample matrix (e.g., blank media or sample diluent). Contact your Avantor representative for more information.
Caution: For research use only.