Specifications
- Description:EasySeed™ Cryptosporidium and Giardia 100
- Size:10 Vials
- Cat. No.:MSPP-ESCG100
Specifications
About this item
EasySeed™ is a global reference standard for Cryptosporidium and Giardia testing.
- Rapid quality control procedure
- Flow cytometer enumerated
- Precise demonstration of testing performance
EasySeed™ contain flow-sorted to contain 100 gamma-irradiated Cryptosporidium parvum (iowa strain) oocysts and Giardia lamblia cysts in approximately 1 ml of saline solution or 100 °C parvum oocysts only. EasySeed™ can simply be added to QC water sample for a precise and consistent demonstration of testing performance.
Gamma-irradiated oocysts and cysts are safe to use and morphologically stable. EasySeed™ tubes are sealed, sterilised and stable for 4 months.