Order Entry
United States
Orders LinkContactUsLinkComponent
Lympholyte®-M Cell Separation Media, Cedarlane

Lympholyte®-M Cell Separation Media, Cedarlane

Marketsource
Supplier:   Cedar Lane
Lympholyte®-M Cell Separation Media, Cedarlane
Supplier:  Cedar Lane
Marketsource
Certificates
Lympholyte®-M Cell Separation Media, Cedarlane

2 Options Available Below

Product Details & Documents

Lympholyte®-M is a density separation medium specifically designed for the isolation of viable lymphocytes from murine lymphoid cell suspensions.

  • Composition: Polysucrose 400 and sodium diatrizoate
  • Density: 1.0875 ±0.0010 g/cm3 at 22 °C
  • pH: 6.9 ±0.3
  • Viability/Purity: Recovery of viable lymphocytes >70%

Lympholyte®-M can be utilized in a simple protocol to eliminate erythrocytes, dead cells and debris from murine spleen, lymph node, thymus and bone marrow suspensions. The resulting cell population demonstrates a high and nonselective recovery of viable lymphocytes that are suitable for use as target cells in cytotoxicity, FACS assays, and in in vivo and in vitro functional studies.

Applications include the removal of dead cells in sequential cytotoxicity studies, the removal of erythrocytes, dead cells and debris from other murine tissue
suspensions including liver and lung, the harvesting of viable cells and removal of dead cells and debris from various clone cell and hybridoma cell lines and the isolation of murine nuclear epidermal cells.

Product Family Options

Product Information

  • Availability
    Price
  • MSPP-CL5030
    Lympholyte®-M cell separation media, sterile liquid, 5×30 ml
    Lympholyte®-M cell separation media, sterile liquid, 5×30 ml
    Log in for availability
    Log in for availability
    Each
    Each - $293.85
    $293.85
    Each
       $293.85
    Minimum order quantity is 1

    Supplier Number: CL5030
    Specifications:
    Marketsource
    Marketsource
  • MSPP-CL5031100
    Lympholyte®-M cell separation media, sterile liquid, 1×100 ml
    Lympholyte®-M cell separation media, sterile liquid, 1×100 ml
    Log in for availability
    Log in for availability
    Each
    Each - $178.46
    $178.46
    Each
       $178.46
    Minimum order quantity is 1

    Supplier Number: CL5031
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource