ContactUsLinkComponent
EasyPlus Density Meters, METTLER TOLEDO®
EasyPlus Density Meters, METTLER TOLEDO®
Supplier:  Mettler Toledo
Certificates
undefined
EasyPlus Density Meters, METTLER TOLEDO®
Supplier:  Mettler Toledo
Specifications
  • Catalog No:
  • 76307-438
  • 76307-440
  • Model:
  • D30
  • D40
  • Repeatability:
  • 0.00005 g/cm3
  • 0.00005 g/cm3
  • Temperature Control:
  • 15…25 °C
  • 15…25 °C
  • W×D×H:
  • 170W×156H×273D mm
  • 170W×156H×273D mm