Sediment Test Cards and Discs, Nelson Jemeson
Catalog # MSPP-7233008
Supplier: NELSON-JAMESON INC MS
CAS Number:
Specifications
- Pack type:500/Box
- Description:Sediment Test Cards and Discs
- Cat. No.:MSPP-7233008
Specifications
About this item
Test cards filter the medium and provide a positive record in one step. They consist of a reporting card with filter disc attached. Accommodate up to 1.25" aperture openings.