Order Entry
United States
ContactUsLinkComponent
IC Anions Standard, Mix 2, Agilent Technologies
IC Anions Standard, Mix 2, Agilent Technologies
Catalog # ULICC-210
Supplier:  Ultra Scientific
Small Business Enterprise
CAS Number:  
undefined
IC Anions Standard, Mix 2, Agilent Technologies
Catalog # ULICC-210
Supplier:  Ultra Scientific
Supplier Number:  ICC-210
Small Business Enterprise
CAS Number:  

Specifications

  • Matrix:
    H₂O
  • Pack type:
    Bottle
  • Description:
    IC Anions Standard
  • Size:
    125 ml
  • Cat. No.:
    ULICC-210
  • Supplier no.:
    ICC-210

Specifications

About this item

Multi component mixture.

  • Solvent: Water (low TOC, 50 ppb)
  • Agency method: EPA 300.1, EPA 300 and EPA 9056A