24 Well Black Flat Bottom Microplates, Sterile, Ibidi
Supplier: Avantor
Certificates
About this item
24 well plates with flat and clear bottom ideal for high throughput applications in cell-based assays.
- Ideal for cell microscopy
- Excellent inner well flatness and whole plate flatness
- Low well-to-well crosstalk in fluorescence microscopy
- Compatible with automation equipment
- Suitable for fluorescence scanners
- High-quality plastic bottom, which is compatible with solvents for staining and fixation and also with immersion oil
- Sterile, single packaging
Used for fluorescence-based imaging of living or fixed cells, high throughput screening (HTS) in cell culture, wound healing experiments with ibidi culture-inserts, large-scale transfection experiments.
Inner well flatness: ±10 µm, whole plate flatness: ±25 µm, coating area per well using 1 ml: 4.30 cm2, well clearance: 1.2 mm, well-to-well distance: 18.9 mm.
Certifications: Meets ANSI/SLAS (SBS) standards.
Frequently Bought Together