Pipet-Aid™ Portable Pipet-Aid, Bellco Glass
Catalog # 101416-104
Supplier: Bellco Glass
Specifications
- Description:Portable Pipet-Aid
- Cat. No.:101416-104
- Supplier no.:1225-85000
Specifications
About this item
Rechargeable self-contained unit that provides two hours of pipetting before recharging is necessary.
- Each button has three speed settings (fast, medium, slow)
Delivery: Includes battery charger, four replacement filters and tissue culture nose piece.