Pipet-Aid™ Pip-Aid Hood Mate, 115 V, Bellco Glass
Catalog # 100484-774
Supplier: Bellco Glass
Specifications
- Description:Pip-Aid Hood Mate
- Cat. No.:100484-774
- Electrical:115 V
- Supplier no.:1225-80300
Specifications
About this item
Coiled vacuum/pressure line prevents contamination inside the hood.
- Delivers precise volumes with exact control
Delivery: Includes mounting bracket for easy installation.