Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS Ultra LabReady™, Solvent Blend tested for UHPLC-MS, Riedel-de Haën™
Catalog # BJ14282-2L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2 L
- Cat. No.:BJ14282-2L
Specifications
Specification Test Results
- Assay of acetic acid:0.093 - 0.107 %
- pH (20 °C):3.2 - 3.5
- Calcium (Ca):Max. 0.2 ppm
- Potassium (K):Max. 0.2 ppm
- Magnesium (Mg):Max. 0.2 ppm
- Sodium (Na):Max. 0.5 ppm
- Transmittance at 254 nm:Min. 99 %
- Transmittance at 230 nm:Min. 75 %
- Transmittance at 210 nm:Min. 20 %
- UHPLC-MS ESI(+) (Reserpine):Max. 10 ppb
- UHPLC-MS ESI(-) (Digoxin):Max. 50 ppb
- Suitability for the UHPLC-MS:complying
- LC-MS application:complying
- HPLC Gradient at 254 nm:Max. 2 mAU
- HPLC Gradient at 210 nm:Max. 20 mAU
Specification Test Results
About this item
:
Density:1.000 g/cm³
Storage Temperature:Ambient
EINECS:231-791-2
:7732-18-5