Water, CHROMASOLV™ LC-MS Ultra tested for UHPLC-MS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Boiling Pt:100 °C (1.013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:EPA Safer chemical solvent (green circle rating); Chem21 solvent selection guide recommended by default and after discussion
Specifications
Specification Test Results
- Non-volatile matter:Max. 1 ppm
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Calcium (Ca):Max. 0.1 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- Copper (Cu):Max. 0.02 ppm
- Iron (Fe):Max. 0.1 ppm
- Potassium (K):Max. 0.1 ppm
- Magnesium (Mg):Max. 0.1 ppm
- Manganese (Mn):Max. 0.02 ppm
- Sodium (Na):Max. 0.1 ppm
- Nickel (Ni):Max. 0.02 ppm
- Lead (Pb):Max. 0.1 ppm
- Tin (Sn):Max. 0.1 ppm
- Zinc (Zn):Max. 0.1 ppm
- Chloride (Cl):Max. 0.01 ppm
- Fluoride (F):Max. 0.01 ppm
- Nitrate (NO₃):Max. 0.1 ppm
- Sulfate (SO₄):Max. 0.1 ppm
- UHPLC-MS ESI(+) (Reserpine):Max. 5 ppb
- UHPLC-MS ESI(-) (Digoxin):Max. 20 ppb
- UHPLC Gradient drift 254 nm:Max. 3 mAU
- UHPLC Gradient peak 254 nm:Max. 0.6 mAU
- UHPLC Gradient drift 210 nm:Max. 8 mAU
- UHPLC Gradient peak 210 nm:Max. 2 mAU
- Transmittance at 230 nm:Min. 99 %
- Transmittance at 200 nm:Min. 95 %
- Fluorescence (chinin) at 254 nm:Max. 1 ppb
- Fluorescence (chinin) at 365 nm:Max. 1 ppb
- Suitability for the UHPLC-MS:complying
Specification Test Results
Related Information
Frequently Bought Together