Order Entry
United States
ContactUsLinkComponent
Tetramethylammonium hydroxide 0.1 M (CH3)4NOH (0.1N) in 2-propanol/methanol volumetric solution, for titration in non-aqueous media, Fluka™
Tetramethylammonium hydroxide 0.1 M (CH3)4NOH (0.1N) in 2-propanol/methanol volumetric solution, for titration in non-aqueous media, Fluka™
Catalog # BJ35436-1L
CAS Number:  
Tetramethylammonium hydroxide 0.1 M (CH3)4NOH (0.1N) in 2-propanol/methanol volumetric solution, for titration in non-aqueous media, Fluka™
Catalog # BJ35436-1L
Supplier Number:  35436-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ35436-1L

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001

Specification Test Results

About this item

 :
 :91.15
Flash Pt:12 °C
Storage Temperature:Ambient
UN:1992
ADR:3,II
 :GHS08
 :GHS06
 :GHS02
CAS Number:75-59-2
Formula:C₄H₁₃NO
MDL Number:MFCD00008280