
tetra-Chloroauric(III) acid trihydrate ≥49.0% (Au basis) ACS, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:16961-25-4
:
Formula:HAuCl₄·3H₂O
MDL Number:MFCD00149904
:393.83 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt:30 °C
Density:3.9 g/cm³
Storage Temperature:Ambient
EINECS:240-948-4
UN:3260
ADR:8,II
:GHS05
Specification Test Results
- Appearance of the substance:complying
- Assay of Au:Min. 49.0 %
- Alkali- and other metals (as SO₄):Max. 0.2 %
- In ether insoluble matter:Max. 0.1 %
Specification Test Results
Related Information
Frequently Bought Together