Order Entry
United States
ContactUsLinkComponent
Tashiro’s indicator solution in methanol, Fluka™
Flammable
Health
Tashiro’s indicator solution in methanol, Fluka™
Catalog # BJ35325-1L
CAS Number:  
Flammable
Health
Tashiro’s indicator solution in methanol, Fluka™
Catalog # BJ35325-1L
Supplier Number:  35325-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    BJ35325-1L

Specifications

Specification Test Results

  • Change range:
    complying

Specification Test Results

About this item

 :
Flash Pt:18 °C
Storage Temperature:Ambient
UN:1230
ADR:3,II
 :GHS02
 :GHS08
MDL Number:MFCD00148563