
Sodium hydroxide, anhydrous ≥98% (by acidimetry), pellets, BioXtra
BJS8045-1KG
:
- Pack type:Plastic bottle
- Size:1 kg
- Cat. No.:BJS8045-1KG
- Supplier no.:S8045-1KG
- Insoluble matter:Max. 0.1 %
- Solution 1 M in H₂O:complying
- Assay (acidimetric):Min. 98.0 %
- Aluminium (Al):Max. 0.0005 %
- Calcium (Ca):Max. 0.0005 %
- Copper (Cu):Max. 0.0005 %
- Iron (Fe):Max. 0.0005 %
- Potassium (K):Max. 0.02 %
- Magnesium (Mg):Max. 0.0005 %
- Phosphorus (P):Max. 0.0005 %
- Lead (Pb):Max. 0.001 %
- Zinc (Zn):Max. 0.0005 %
- Chloride (Cl):Max. 0.005 %
- Sulfate (SO₄):Max. 0.05 %
- Form:pellets (anhydrous)
:
:40 g/mol
:1.390 °C (1.013 hPa)
:319 °C
:2.13 g/cm³ (20 °C)
:Ambient
:215-185-5
:1823
:8,II
:GHS05
:1310-73-2
:NaOH
:MFCD00003548