Sodium hydroxide 0.111 M NaOH (0.111N) volumetric solution, for milk acidity analysis according to Dornic, Fluka™
Catalog # BJ34283-1L
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ34283-1L
Specifications
Specification Test Results
- Vapor Pressure:3 mmHg ( 37 °C)
- Factor:0.999 - 1.001
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:100 °C (1.013 hPa)
Density:1.000 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-791-2
UN:1824
ADR:8,III
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548