Order Entry
United States
ContactUsLinkComponent
 
 
Sodium hydroxide ≥99.99% (trace metals basis), pellets, Semiconductor grade
 
 
Sodium hydroxide ≥99.99% (trace metals basis), pellets, Semiconductor grade
 

 

  • Vapor Pressure:
    3 mmHg ( 37 °C)
  • Appearance of the substance:
    complying
  • Assay:
    Min. 95.0 %
  • Assay of Na₂CO₃:
    Max. 1.0 %
  • Boron (B):
    Max. 10 ppm
  • Cesium (Cs):
    Max. 20 ppm
  • Potassium (K):
    Max. 100 ppm
  • Lithium (Li):
    Max. 10 ppm
  • Rubidium (Rb):
    Max. 10 ppm

 

Frequently Bought Together