Sodium bromide ≥99%, Reagent Grade
Catalog # BJ220345-12KG
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Drum with inliner
- Size:12 kg
- Cat. No.:BJ220345-12KG
Specifications
Specification Test Results
- Assay:99.0 - 101.0 %
- Assay of Br:76.8 - 78.4 %
Specification Test Results
About this item
:
:102.89 g/mol
Boiling Pt:1.393 °C (1.013 hPa)
Melting Pt:750 °C
Density:3.2 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-599-9
CAS Number:7647-15-6
Formula:NaBr
MDL Number:MFCD00003475