Pumice stone, granular 4-6 mm, Fluka™
Catalog # BJ51753-1KG
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:1 kg
- Cat. No.:BJ51753-1KG
Specifications
Specification Test Results
- Grain Size 5-8 Mm:Min. 85 %
- Loss on ignition (800°C):Max. 4 %
Specification Test Results
About this item
:
Storage Temperature:Ambient
CAS Number:1332-09-8
MDL Number:MFCD00148147