Order Entry
United States
ContactUsLinkComponent
Propylene carbonate ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Propylene carbonate ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # BJ414220-1L
Supplier:  Avantor
CAS Number:  
Propylene carbonate ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # BJ414220-1L
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Environmentally Preferable:
  • Cat. No.:
    BJ414220-1L

Specifications

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.65 %
  • Water (Karl Fischer):
    Max. 0.04 %
  • Absorbance at 400 nm:
    Max. 0.01
  • Absorbance at 375 nm:
    Max. 0.01
  • Absorbance at 350 nm:
    Max. 0.05
  • Absorbance at 300 nm:
    Max. 0.30
  • Absorbance 280 nm:
    Max. 0.50
  • Absorbance at 235 nm:
    Max. 1.0
  • Refractive Index:
    n20/D 1.421 (lit.)
  • Vapor Pressure:
    0.98 mmHg ( 50 °C)

Specification Test Results

About this item

Synonyms: Propylene glycol carbonate, 4-Methyl-1,3-dioxolan-2-one, Carbonic acid propylene glycol ester
 :
 :102.09 g/mol
Boiling Pt:242 °C (1.013 hPa)
EINECS:203-572-1
 :GHS07
Melting Pt:-48.8
Storage Temperature:Ambient
CAS Number:108-32-7
Formula:C₄H₆O₃
MDL Number:MFCD00005385
Density:1.205 g/cm³ (20 °C)
Flash Pt:135 °C