Potassium dihydrogen phosphate ≥99.995% (metals basis), TraceSELECT™ for trace analysis, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt:253 °C
Density:2.34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-913-4
Specification Test Results
- Barium (Ba):Max. 1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.005 ppm
- Cobalt (Co):Max. 0.01 ppm
- Chromium (Cr):Max. 0.5 ppm
- Copper (Cu):Max. 0.005 ppm
- Iron (Fe):Max. 0.1 ppm
- Iridium (Ir):Max. 0.5 ppm
- Magnesium (Mg):Max. 0.5 ppm
- Manganese (Mn):Max. 0.01 ppm
- Sodium (Na):Max. 20 ppm
- Nickel (Ni):Max. 0.01 ppm
- Lead (Pb):Max. 0.005 ppm
- Palladium (Pd):Max. 0.5 ppm
- Platinum (Pt):Max. 0.5 ppm
- Rhodium (Rh):Max. 0.5 ppm
- Ruthenium (Ru):Max. 0.5 ppm
- Samarium (Sm):Max. 0.01 ppm
- Strontium (Sr):Max. 0.5 ppm
- Thallium (Tl):Max. 0.01 ppm
- Vanadium (V):Max. 0.1 ppm
- Zinc (Zn):Max. 0.005 ppm
- Cerium (Ce):Max. 0.01 ppm
- Europium (Eu):Max. 0.01 ppm
- Lanthanum (La):Max. 0.01 ppm
- Yttrium (Y):Max. 0.01 ppm
- Ytterbium (Yb):Max. 0.01 ppm
- Arsenic (As):Max. 0.01 ppm
- Mercury (Hg):Max. 5 ppb
- Antimony (Sb):Max. 0.5 ppm
- Selenium (Se):Max. 0.01 ppm
- Chloride (Cl):Max. 5 ppm
- Sulfate (SO₄):Max. 30 ppm
- Total N:Max. 10 ppm
Specification Test Results
Frequently Bought Together