Order Entry
United States
ContactUsLinkComponent
Oxalic acid concentrate for dilution to 1L, 0.005 M (COOH)2 (0.01N) volumetric solution, Fluka™
Oxalic acid concentrate for dilution to 1L, 0.005 M (COOH)2 (0.01N) volumetric solution, Fluka™
Catalog # BJ38255-1EA
CAS Number:  
Oxalic acid concentrate for dilution to 1L, 0.005 M (COOH)2 (0.01N) volumetric solution, Fluka™
Catalog # BJ38255-1EA
Supplier Number:  38255-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Size:
    1 Each
  • Cat. No.:
    BJ38255-1EA

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :90.04 g/mol
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573