Orthoboric acid 99.5-100.5%, powder, Puriss., meets analytical specification of BP, Ph. Eur., NF, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
Synonyms: Boron trihydroxide, Orthoboric acid, Boron hydroxide, Boric acid, Boracic acid
CAS Number:10043-35-3
:
Formula:B(OH)₃
MDL Number:MFCD00011337
Boiling Pt:185
Melting Pt:160
Density:1.510 g/cm³ (20 °C)
Molecular Weight:61.83 g/mol
Merck Index:13,01326
EINECS:233-139-2
:GHS08
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:99.5 - 100.5 %
- Loss on drying (over silicagel, 5h):Max. 0.5 %
- pH (3,3 %, 20 °C):3.8 - 4.8
- Arsenic (As):Max. 0.0005 %
- Iron (Fe):Max. 0.0005 %
- Heavy metals (as Pb):Max. 0.001 %
- Sulfate (SO₄):Max. 0.04 %
- Organic matter:complying
- Residual solvents:complying
- Solubility in C₂H₅OH:complying
- Appearance of the solution:complying
- Completeness of the solution:complying
Specification Test Results
Frequently Bought Together