
Nitric acid 2 M HNO3 (2N) volumetric solution, Fluka™
BJ35278-1L
:
- Pack type:Plastic bottle
- Size:1 L
- Cat. No.:BJ35278-1L
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
:
:63.01
:100 °C
:1.065 g/cm³ (20 °C)
:Ambient
:2031
:8,II
:GHS05
:7697-37-2
:HNO₃
:MFCD00011349