Molecular sieve A4 (0.4 nm, 4 Å), beads 8 - 12 mesh, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:70955-01-0
:1318-02-1
MDL Number:MFCD03457537
Melting Pt:400 °C
Density:2.0 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Particle Size:8-12 mesh
- Grain size 8 - 12 mesh:Min. 95 %
Specification Test Results
Related Information
Frequently Bought Together