Order Entry
United States
ContactUsLinkComponent
Molecular sieve 3A (0.3 nm, 3 Å), beads 8-12 mesh, Fluka™
Molecular sieve 3A (0.3 nm, 3 Å), beads 8-12 mesh, Fluka™
Certificates
Molecular sieve 3A (0.3 nm, 3 Å), beads 8-12 mesh, Fluka™
Certificates

Specification Test Results

  • Form:
    beads
  • Grain size 8 - 12 mesh:
    Min. 95 %
  • Pore Size (Ång Ström):
    3

Specification Test Results

Related Information

Frequently Bought Together