Magnesium chloride hexahydrate 99-101%, Puriss., meets analytical specification of BP, FCC, Ph. Eur., E511 (max. 0.0001% Al), Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:7791-18-6
:
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781
:203.3 g/mol
Melting Pt:117 °C (–H₂O of crystall.)
Density:1.57 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Assay:99.0 - 101.0 %
- Water (Karl Fischer):51.0 - 55.0 %
- Free acid (as HCl):Max. 0.02 %
- Free alkali (as MgO):Max. 0.01 %
- Acidity or alkalinity:complying
- Ammonium (NH₄):Max. 50 ppm
- Aluminium (Al):Max. 1 ppm
- Arsenic (As):Max. 2 ppm
- Barium (Ba):Max. 0.002 %
- Calcium (Ca):Max. 0.1 %
- Iron (Fe):Max. 5 ppm
- Mercury (Hg):Max. 1 ppm
- Potassium (K):Max. 500 ppm
- Sodium (Na):Max. 0.5 %
- Lead (Pb):Max. 2 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Bromide (Br):Max. 500 ppm
- Sulfate (SO₄):Max. 100 ppm
- Appearance of the solution:complying
- Residual solvents:complying
Specification Test Results
Frequently Bought Together