

Lithium hydroxide monohydrate ≥98.0% ACS, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:1310-66-3
:
Formula:LiOH·1H₂O
MDL Number:MFCD00149772
:41.96 g/mol
Melting Pt:423 °C
Density:1.830 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2680
ADR:8,II
:GHS07
:GHS05
Specification Test Results
- Assay:Min. 98.0 %
- Assay of Li₂CO₃:Max. 2.0 %
- Water insoluble matter:Max. 0.01 %
- Iron (Fe):Max. 0.002 %
- Heavy metals (ICP) ACS:Max. 0.002 %
- Chloride (Cl):Max. 0.01 %
- Sulfate (SO₄):Max. 0.05 %
Specification Test Results
Frequently Bought Together