Order Entry
United States
ContactUsLinkComponent
Lithium hydroxide monohydrate ≥98.5%, Purum, Fluka™
Irritant
Corrosive
Lithium hydroxide monohydrate ≥98.5%, Purum, Fluka™
Certificates
Irritant
Corrosive
Lithium hydroxide monohydrate ≥98.5%, Purum, Fluka™

Specification Test Results

  • Assay:
    Min. 98.5 %
  • Assay of Li₂CO₃:
    Max. 1 %
  • Calcium (Ca):
    Max. 0.05 %
  • Iron (Fe):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.02 %
  • Sodium (Na):
    Max. 0.02 %
  • Chloride (Cl):
    Max. 0.05 %
  • Sulfate (SO₄):
    Max. 0.05 %

Specification Test Results

Related Information

Frequently Bought Together