
Lithium chloride, anhydrous ≥99% ACS, free-flowing, Fluka™
Catalog # BJ746460-500G
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:6 x 500 g
- Cat. No.:BJ746460-500G
Specifications
Specification Test Results
- Assay:Min. 99.0 %
- Water insoluble matter:Max. 0.01 %
- Loss on drying (105°C, 2 h):Max. 1.0 %
- Titrable Base:Max. 0.008 meq/g
- Barium (Ba):Max. 0.003 %
- Calcium (Ca):Max. 0.01 %
- Iron (Fe):Max. 0.001 %
- Potassium (K):Max. 0.01 %
- Sodium (Na):Max. 0.20 %
- Heavy metals (ICP) ACS:Max. 0.002 %
- Nitrate (NO₃):Max. 0.001 %
- Sulfate (SO₄):Max. 0.01 %
Specification Test Results
About this item
:
:42.39 g/mol
Boiling Pt:1.360 °C (1.013 hPa)
Melting Pt:605 °C
Density:2.07 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-212-3
:GHS07
CAS Number:7447-41-8
Formula:LiCl
MDL Number:MFCD00011078