Order Entry
United States
Orders LinkContactUsLinkComponent
Iodine ≥99.8%, solid ACS, Fluka™

Iodine ≥99.8%, solid ACS, Fluka™

Iodine ≥99.8%, solid ACS, Fluka™
Certificates

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Honeywell ACS solvents for general laboratory use are:

  • A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested tmeet ACS general use specifications
  • CAS Number:7553-56-2
  • Formula:I₂
  • MDL Number:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Melting Pt:113.5 °C
  • Density:4.930 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-442-4
  • UN:3495
  • ADR:8,III
Specifications for Product Family

Specification Test Results

  • Assay:
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.01 %
  • Bromine and chlorine (as chloride):
    Max. 0.005 %
  • Form:
    Solid
  • Vapor Pressure:
    0.31 mmHg ( 25 °C)
  • Resistivity:
    1.3E15 μΩ-cm

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $142.09
    $142.09
    Each (100g)
       $142.09

    Supplier Number: 207772-100G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    BJ207772-100G
  • Glass bottle
    1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $761.15
    $761.15
    Each (1kg)
       $761.15

    Supplier Number: 207772-1KG
    Specifications:
    More
    Specifications
    Cat. No.
    BJ207772-1KG
  • Glass bottle
    2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $1,708.05
    $1,708.05
    Each (2,500g)
       $1,708.05

    Supplier Number: 207772-2.5KG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    BJ207772-2.5KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.