Disodium hydrogen phosphate dodecahydrate 98.5-101% (dried basis), meets analytical specification of BP, Ph. Eur., E339, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
Synonyms: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate
CAS Number:10039-32-4
:
Formula:Na₂HPO₄·12H₂O
MDL Number:MFCD00149181
:358.14 g/mol
Melting Pt:34 °C
Density:1.52 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-448-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:98.5 - 102.5 %
- Assay (calculated to the dried substance):98.5 - 101 %
- Assay of NaH₂PO₄ (calculated to dried substances):Max. 1.0 %
- Water (Karl Fischer):57.0 - 61.0 %
- Loss on drying (130°C):Max. 61.0 %
- Water insoluble matter:Max. 0.2 %
- pH (1 %,20°C):8.4 - 9.6
- Arsenic (As):Max. 1 ppm
- Cadmium (Cd):Max. 1 ppm
- Copper (Cu):Max. 0.002 %
- Iron (Fe):Max. 10 ppm
- Mercury (Hg):Max. 1 ppm
- Potassium (K):Max. 0.005 %
- Lead (Pb):Max. 1 ppm
- Zinc (Zn):Max. 0.001 %
- Heavy metals (as Pb):Max. 10 ppm
- Chloride (Cl):Max. 10 ppm
- Fluoride (F):Max. 10 ppm
- Sulfate (SO₄):Max. 100 ppm
- Appearance of the solution:complying
- Reducing impurities:complying
- Residual solvents:complying
Specification Test Results
Related Information
Frequently Bought Together