
Bromine (bromide-bromate) solution concentrate for dilution to 1L, 0.05 M Br2 volumetric solution, Fluka™
Catalog # BJ38040-1EA
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Size:1 Each
- Cat. No.:BJ38040-1EA
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
:GHS08
Storage Temperature:Ambient
MDL Number:MFCD00143870