Order Entry
United States
ContactUsLinkComponent
 
Barium sulfate 97.0-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
 
Barium sulfate 97.0-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., Fluka™

 

  • Assay:
    97.5 - 100.5 %
  • Loss on ignition (600°C):
    Max. 2.0 %
  • Soluble in acid:
    Max. 0.3 %
  • Soluble Ba-salts:
    complying
  • Acid. or alk. react. impurities:
    complying
  • Heavy metals (as Pb):
    max. 0.001 %
  • Oxidizable S compounds:
    complying
  • Residual solvents:
    complying

 

Frequently Bought Together