


Ammonium persulfate (APS) ≥98%, Puriss. p.a. ACS, Reag. Ph. Eur., Fluka™
Catalog # BJ31117-5KG
Supplier: Honeywell Research Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:5 kg
- Cat. No.:BJ31117-5KG
- Supplier no.:31117-5KG
Specifications
Specification Test Results
- Assay:Min. 98 %
- Water insoluble matter:Max. 0.005 %
- Residue on ignition (SO₄):Max. 0.05 %
- Titrable Acid:Max. 0.04 meq/g
- Iron (Fe):Max. 0.001 %
- Manganese (Mn):Max. 0.00005 %
- Heavy metals (as Pb):Max. 0.001 %
- Chloride, Chlorate (as Cl):Max. 0.001 %
- Chloride (Cl):Max. 0.0005 %
Specification Test Results
About this item
:
:228.2 g/mol
Melting Pt:120 °C
Density:1.98 g/cm³
Storage Temperature:Ambient
UN:1444
ADR:5.1,III
:GHS07
:GHS08
:GHS03
CAS Number:7727-54-0
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390