Order Entry
United States
ContactUsLinkComponent
Ammonium iron (III) sulfate dodecahydrate, slightly violet 99.0-102.0%, crystalline, Puriss. p.a. ISO, ACS, Reag. Ph. Eur., Fluka™
Ammonium iron (III) sulfate dodecahydrate, slightly violet 99.0-102.0%, crystalline, Puriss. p.a. ISO, ACS, Reag. Ph. Eur., Fluka™
Certificates
Ammonium iron (III) sulfate dodecahydrate, slightly violet 99.0-102.0%, crystalline, Puriss. p.a. ISO, ACS, Reag. Ph. Eur., Fluka™

Specification Test Results

  • Appearance:
    complying
  • Assay:
    99.0 - 102.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • Insoluble in HCl:
    Max. 0.01 %
  • Calcium (Ca):
    Max. 0.002 %
  • Copper (Cu):
    Max. 0.001 %
  • Iron (Fe 2⁺):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.001 %
  • Manganese (Mn):
    Max. 0.005 %
  • Sodium (Na):
    Max. 0.01 %
  • Lead (Pb):
    Max. 0.0005 %
  • Zinc (Zn):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.0005 %
  • Nitrate (NO₃):
    Max. 0.01 %
  • Phosphate (PO₄):
    Max. 0.002 %
  • Solution 10 g to 100 mL (H₂O):
    complying
  • Form:
    crystalline

Specification Test Results

Related Information

Frequently Bought Together