Order Entry
United States
ContactUsLinkComponent
Ammonium heptamolybdate tetrahydrate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Ammonium heptamolybdate tetrahydrate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Certificates
Ammonium heptamolybdate tetrahydrate ≥99.0% (by titrimetric analysis), Puriss. p.a. ACS, Fluka™
Certificates

Specification Test Results

  • Assay:
    99.4 - 101.8 %
  • Assay (as MoO₃):
    81.0 - 83.0 %
  • Water insoluble matter:
    Max. 50 ppm
  • Calcium (Ca):
    Max. 10 ppm
  • Cadmium (Cd):
    Max. 5 ppm
  • Cobalt (Co):
    Max. 5 ppm
  • Chromium (Cr):
    Max. 5 ppm
  • Copper (Cu):
    Max. 5 ppm
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 50 ppm
  • Magnesium (Mg):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 5 ppm
  • Sodium (Na):
    Max. 50 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Zinc (Zn):
    Max. 5 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Heavy metals (ICP) ACS:
    Max. 0.001 %
  • Chloride (Cl):
    Max. 10 ppm
  • Nitrate (NO₃):
    Max. 20 ppm
  • Phosphate (PO₄):
    Max. 5 ppm
  • Phosphate, Arsenate, Silicate (as SiO₂):
    Max. 10 ppm
  • Sulfate (SO₄):
    Max. 100 ppm

Specification Test Results

Frequently Bought Together