ContactUsLinkComponent
CE Softwall Cleaner™, Cleaning Mop, Vileda Professional®
CE Softwall Cleaner™, Cleaning Mop, Vileda Professional®
Catalog # 76318-338
undefined
CE Softwall Cleaner™, Cleaning Mop, Vileda Professional®
Catalog # 76318-338
Supplier Number:  162232
Specifications
  • Description:
    CE Softwall Cleaner™
  • Material:
    Electro-Polished Stainless Steel and Aluminum
  • Cat. No.:
    76318-338
Specifications
About this item
CE Softwall Cleaner™ is designed to clean soft wall curtains separating zones within cleanrooms. Robust construction and 'shotgun pump' action design means higher pressure can be applied during cleaning and easily adjusted as needed.
  • Completely autoclavable
  • Telescopic handle reaches to 9"
  • Unique design means higher pressure can be applied during cleaning and adjusted as necessary
  • Also allows full contact on curtain from ceiling to floor
Customized tools for special surfaces. The easiest, most effective way to clean pipes, soft walls curtains, tanks and other hard to reach areas in Controlled Environments.
A new line of specialty above-the-floor tools by Vileda Professional®. This line is ideal for all surfaces that are hard or difficult to reach, clean, and disinfect. Specialty tools use Controlled Environment compatible disposable microfiber sleeves that are already validated materials and are an extension of a current CE floor cleaning system.
Certifications: Meet CE requirements up to ISO 4-9/GMP A/B, C, D.