ContactUsLinkComponent
Impregnated Gauze Dressing, Dukal™ Corporation
Catalog # 76321-386
Supplier:  VWR International
Impregnated Gauze Dressing, Dukal™ Corporation
Catalog # 76321-386
Supplier:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Sterility:
    Sterile
  • Color:
    White/Blue
  • Description:
    Petrolatum Dressing
  • Dimensions:
    3×9"
  • Environmentally Preferable:
  • Cat. No.:
    76321-386
Specifications
About this item
Impregnated gauze dressing provide soothing non-adherent protection with more convenience and less mess.
  • Non-adherent petrolatum
  • Clings and conforms to all body contours
  • Non-toxic, non-irritating, guaranteed sterile
  • Latex-free
Dressings can be used as an initial layer in surgical dressings, on skin graft sites, for treatment of first degree burns or abrasions, and as wound packing.