ContactUsLinkComponent
Glucose Uptake-Glo™ Assay, Promega
Glucose Uptake-Glo™ Assay, Promega
  76201-554
 :  
undefined
Glucose Uptake-Glo™ Assay, Promega
  76201-554
 :  J1341
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Glucose Uptake-Glo™ Assay Kit
  • Size:
    1 Kit
  • Environmentally Preferable:
  • Cat. No.:
    76201-554
A simple, non-radioactive assay used to measure glucose uptake inside cells.
  • Homogeneous protocol
  • Achieve sensitivity with broad linearity
  • Reliable and reproducible results
Glucose metabolism is a key process in many organisms. A lack of insulin-stimulated glucose uptake is associated with type 2 diabetes, while high glucose uptake is a sign of the high glycolytic rates associated with cancer. Measuring glucose uptake can determine the effects of various treatments for diabetes and cancer.
The gold standard method of assaying glucose uptake relies on detection of radio-labelled glucose analogs. Alternative colorimetric and fluorometric glucose uptake assays often lack the sensitivity and robustness needed to reliably measure glucose uptake in cells.
 : The system contains sufficient reagents to perform 50 reactions in 96-well plates (50 µl of sample, 25 µl of stop buffer, 25 µl of neutralization buffer, 100 µl of 2DG6P detection reagent). It includes luciferase reagent (5 ml), stop buffer (15 ml), neutralization buffer (15 ml), NADP+ (20mM) (50 μl), glucose-6-phosphate dehydrogenase (G6PDH) (125 μl), reductase (25 μl), reductase substrate (55 μl), 2-deoxyglucose (2DG, 100mM) (250 μl), 2DG6P standard (1mM) (50 μl). Store the kit at –65 °C.
 : For research use only. Not for use in diagnostic procedures.