Order Entry
United States
ContactUsLinkComponent
Lotus Tetragonolobus Lectin, Fluorescein
Lotus Tetragonolobus Lectin, Fluorescein
Catalog # 101098-026
CAS Number:  
undefined
Lotus Tetragonolobus Lectin, Fluorescein
Catalog # 101098-026
Supplier Number:  FL-1321-2
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Fluorescein
  • Size:
    2 mg
  • Protein/Peptide Name:
    Lotus Tetragonolobus Lectin
  • Cat. No.:
    101098-026
  • Supplier no.:
    FL-1321

Specifications

About this item

Lotus tetragonolobus lectin, is a family of closely related glycoproteins that appear to have similar specificities toward α-linked L-fucose containing oligosaccharides.

  • Supplied essentially free of unconjugated fluorochromes
  • Inhibiting/eluting sugar: 50 to 100 mM L-fucose

Fluorescein labeled Lotus tetragonolobus lectin has an appropriate number of fluorochromes bound to provide the optimum staining characteristics for this lectin. The excitation maximum is at 495 nm and the emission maximum is at 515 nm.