Order Entry
United States
ContactUsLinkComponent
High Capacity Magne® Streptavidin Beads, Promega
High Capacity Magne® Streptavidin Beads, Promega
  76201-362
undefined
High Capacity Magne® Streptavidin Beads, Promega
  76201-362
 :  V7820
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Concentration:
    20% slurry in 10mM phosphate buffer + 0.02% sodium azide
  • Description:
    High capacity Magne® streptavidin beads
  • Size:
    3 ml
  • Particle size:
    30 - 50 μm
  • Binding Capacity:
    ≥9 mg of goat anti-human biotinylated IgG/ml of settled resin or ≥1.8 mg of goat anti-human biotinylated IgG/ml of bead slurry
  • Cat. No.:
    76201-362

 

 

High Capacity Magne® Streptavidin Beads are magnetic affinity beads with high specificity and high capacity for binding biotinylated antibodies and proteins. The magnetic beads are composed of iron oxide encapsulated by macroporous cellulose, which results in beads that have low nonspecific binding.

  • Ideal for use with complex biological samples
  • Excellent magnetic properties for rapid and efficient capture
  • Capture process can be automated for high throughput and scaled up to handle a variety of sample volumes

These beads are used for a variety of applications including pharmacokinetics studies of therapeutic antibodies. The high capacity of the beads enables enrichment of antibodies over a wide concentration range using a small amount of beads.

The affinity of streptavidin for biotin (Kd = 10⁻¹⁵ M) is one of the strongest, most stable interactions in biology; this interaction cannot be reversed under nondenaturing conditions. Hence, it is possible to perform extensive washing to remove nonspecifically bound protein and elute therapeutic antibodies without eluting the biotinylated component, thereby improving the detection limit of the antibody.

 : Store the beads at 2 to 10 °C. Do not freeze. Do not allow beads to dry during storage or use. Do not reuse the beads.