Order Entry
United States
Orders LinkContactUsLinkComponent
Protein Loading Buffer, 4X

Protein Loading Buffer, 4X

Protein loading buffer (4X), 15 ml
Catalog # 102673-500
Marketsource
Supplier:   Li-Cor
Supplier #:  928-40004
Protein Loading Buffer, 4X
Catalog # 102673-500
Supplier:  Li-Cor
Marketsource
CAS Number:  
Protein Loading Buffer, 4X

Protein loading buffer (4X), 15 ml

CAS Number:  
Price
$55.38
Log in for availability
Each (15ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Protein loading buffer (4X) is used as a loading buffer for protein gel electrophoresis.

This buffer is optimized for use with SDS-page and Tris-glycine-SDS running buffer.

It contains Tris-HCl (pH 6.8) 125 mM, glycerol 50%, SDS 4%, orange G 0.2% (w/v).

Specifications

Specifications

  • Description:
    Protein loading buffer (4X)
  • Size:
    15 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.