Orders LinkContactUsLinkComponent
Human Serum, Pooled Complement, Innovative Research
Human Serum, Pooled Complement, Innovative Research
Human Serum, Pooled Complement, 50 ml
Catalog # MSPP-IPLA-CSER
Marketsource
Supplier #:  ICSER50ML
Human Serum, Pooled Complement, Innovative Research
Catalog # MSPP-IPLA-CSER
Marketsource
Human Serum, Pooled Complement, Innovative Research
Human Serum, Pooled Complement, 50 ml
Price
$494.52
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Pooled human complement serum is a complex mixture of serum proteins which are activated sequentially when antibodies in the blood of an immune animal interact with the corresponding antigens.
This serum is collected at an FDA-licensed commercial donor center/facility within the United States. Each unit is tested and found negative for HBsAg, HCV, HIV-1, HIV-2, HIV-1Ag or HIV 1-NAT, ALT, and syphilis by FDA-approved methods.
Caution: This product is for in vitro research use only, is not to be consumed by animals or humans, and should be handled with general precaution as if capable of spreading infectious disease.
Specifications
  • Description:
    Human Serum, Pooled Complement
  • Size:
    50 ml