West Nile Virus Antibody Test Kit, ELISA Kit, Inbios International
MSPP-WNEQM-1A
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Primary antibody reactivity:Horse
- Target protein:IgM
- Description:West Nile Virus Antibody Test Kit
- Size:Kit
- Sample Type:Serum
- Shelf Life:12 Months
- Storage Temperature:2...8 °C
- Cat. No.:MSPP-WNEQM-1A
The West Nile Virus Antibody Test Kit, ELISA is an ELISA assay system for the detection of IgM antibodies in equine serum to WNV-derived recombinant antigen.
: USDA Licensed.
: The kit includes one 96 well strippable plate with all necessary reagents and controls.