ContactUsLinkComponent
Multi-OSMETTE™ Auto-Sampling Turntable Osmometer, Model 2430, Precision System
Multi-OSMETTE™ Auto-Sampling Turntable Osmometer, Model 2430, Precision System
Catalog # MSPP-2430
Multi-OSMETTE™ Auto-Sampling Turntable Osmometer, Model 2430, Precision System
Catalog # MSPP-2430
Supplier Number:  2430
Specifications
  • Range:
    0 - 2000 mOsm/kg
  • Description:
    Multi-OSMETTE™ Auto-sampling turntable osmometer
  • Electrical:
    115 V, 50/60 Hz
  • Sample Volume:
    30 µl
  • Test Time:
    50 - 70 seconds
  • Cat. No.:
    MSPP-2430
Specifications
About this item
Multi-OSMETTE™ auto-sampling turntable osmometer is a laboratory model freezing point osmometer for multiple samples.
  • Works at lowest operating costs, with high accuracy
  • High capacity 24-position turntable, fast 50 to 70 seconds per reading
  • Self identifying, fully automatic calibration
  • Flexible calibration using from 2 to 5 calibration points
  • Built-in ink based printer ranges 0 to 2000 mOsm/kg
  • No copying necessary for records retention
  • Shipped assembled and ready to use
Delivery: Includes instruction manual, printer paper, printer ribbon, sample carousel, evaporation shield, 30 µl sample tubes, one pack of CON-TROL 290 ampules.