Type 17 Short Micro Cuvettes with PTFE Cover, FireFlySci
Supplier: FIREFLYSCI, INC.
Certificates
About this item
The FireflySci Type 17 Short Micro Cuvettes are the quick and cost-effective solution with a height of 25 mm.
- Highly accurate results, only need to test sample once
- Made by hand, ensuring high quality optical polishing with no defects
- Each cell goes through a vigorous inspection process before leaving our factory
- Fire-fused to ensure that cuvettes are robust and durable
- Comes in several different materials: (UV Quartz and IR Quartz)
- Available in 10 mm lightpath
- Two windows optically polished
Accurate and of high quality, Firefly cuvettes are made with the premium quality quartz and glass material available. Excellent transmission values.
Frequently Bought Together