Specifications
- Description:96S super magnet plate
- Cat. No.:MSPP-A001322
- Supplier no.:A001322
Specifications
About this item
96 NdFeB ring magnets, grade N48 are designed for use with the SPRI magnetic bead chemistry. Used widely in laboratories and in variety of magnetic bead protocols.
- Recommended reaction volume up to 1000 µl
- Greatly increased microplate compatibility
- Edge effects are minimized because each well in the plate has its own magnet
- More magnetic material per well makes for faster separations
It can be used with 96-well microplates (shallow and half-deep), round-bottom and V-bottom and also with 96-well PCR plates and strips. Integrated spring cushion technology for complete liquid removal without tip occlusion and gripper grooves on the long sides for robotic access.