

Hanus solution (Iodine bromine) 0.1 M IBr volumetric solution, for determination of iodine value, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
:7789-33-5
Formula:IBr
MDL Number:MFCD00011353
Density:1.06 g/cm³
Flash Pt:40 °C
UN:2789
ADR:8,II
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Assay:0.0985 - 0.1014 mol/l
Specification Test Results
Frequently Bought Together